Assay Details
| Gene Symbol: Gm16576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045069.1 | 3 - 4 | 518 | 60 | ||
| GenBank mRNA | AK143676.1 | - | 3 - 4 | 518 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Sad1 and UNC84 domain containing 2 | Sun2 |
B230369L08Rik, C030011B15, Unc84b |
-2.0kb | Anti-sense | Chr. 15 - 79724068 - 79742536 on NCBI Build GRCm38 |
| Downstream |
dynein, axonemal, light chain 4 |
Dnal4 |
D15Ertd424e, Dnalc4 |
4.1kb | Anti-sense | Chr. 15 - 79761449 - 79774469 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16576
Genomic Map