Assay Details
| Gene Symbol: Gm31452 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143700.1 | - | 1 - 2 | 183 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 3830408C21 gene | 3830408C21Rik | -30.2kb | Sense | Chr. 13 - 107022570 - 107033518 on NCBI Build GRCm38 | |
| Downstream |
apolipoprotein O, pseudogene |
Apoo-ps |
Apoo, EG621156 |
314.9kb | Anti-sense | Chr. 13 - 107414163 - 107414767 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31452
Genomic Map