Assay Details
| Gene Symbol: Gm12596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105031.1 | 1 - 2 | 32 | 77 | ||
| GenBank mRNA | AK143878.1 | - | 1 - 2 | 33 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UDP-glucose ceramide glucosyltransferase | Ugcg |
AU043821, C80537, Epcs21, GlcT-1, Ugcgl |
-37.2kb | Sense | Chr. 4 - 59189204 - 59222833 on NCBI Build GRCm38 |
| Downstream | sushi domain containing 1 | Susd1 |
A530080P10, Gm12528, OTTMUSG00000007332 |
53.5kb | Anti-sense | Chr. 4 - 59314202 - 59439289 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm12596
Genomic Map