Assay Details
| Gene Symbol: Gm19950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143883.1 | - | 1 - 2 | 1312 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 39079 |
Gm39079 | -6.1kb | Sense | Chr. 7 - 119610690 - 119620433 on NCBI Build GRCm38 | |
| Downstream | acyl-CoA synthetase medium-chain family member 4 | Acsm4 |
O-MACS, OMACS |
59.0kb | Anti-sense | Chr. 7 - 119690026 - 119714566 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm19950
Genomic Map