Assay Details
| Gene Symbol: Gm19950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143933.1 | - | 314 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 39079 |
Gm39079 | Sense | Chr. 7 - 119610690 - 119620433 on NCBI Build GRCm38 | ||
| Downstream | acyl-CoA synthetase medium-chain family member 4 | Acsm4 |
O-MACS, OMACS |
58.7kb | Anti-sense | Chr. 7 - 119690026 - 119714566 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm19950
Genomic Map