Assay Details
Gene Symbol: Gm16833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045754.1 | 2 - 3 | 250 | 80 | ||
GenBank mRNA | AK143934.1 | - | 2 - 3 | 250 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | progesterone receptor | Pgr |
9930019P03Rik, BB114106, ENSMUSG00000074510, NR3C3, PR, PR-A, PR-B |
-268.0kb | Sense | Chr. 9 - 8894736 - 8968611 on NCBI Build GRCm38 |
Downstream | contactin 5 | Cntn5 |
6720426O10Rik, A830025P08Rik, Gm507, NB-2 |
335.4kb | Anti-sense | Chr. 9 - 9582665 - 10904775 on NCBI Build GRCm38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK143934.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
