Assay Details
| Gene Symbol: Gm21691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143973.1 | - | 2 - 3 | 656 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
myosin, light polypeptide kinase |
Mylk |
9530072E15Rik, A930019C19Rik, AW489456, KRP, MLCK108, MLCK210, Mlck, nmMlck, smMLCK |
Anti-sense | Chr. 16 - 34745199 - 35002436 on NCBI Build GRCm38 | |
| Downstream | 3-hydroxyacyl-CoA dehydratase 2 | Hacd2 |
6330408J20Rik, AI255777, AI481689, Hcad2, Ptplb |
18.2kb | Anti-sense | Chr. 16 - 35022421 - 35109190 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm21691
Genomic Map