Assay Details
| Gene Symbol: Gm16712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108021.1 | 2 - 3 | 387 | 63 | ||
| GenBank mRNA | AK146869.1 | - | 2 - 3 | 368 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Epstein-Barr virus induced gene 3 | Ebi3 |
EBI-3, IL-27 |
Anti-sense | Chr. 17 - 55952623 - 55957022 on NCBI Build GRCm38 | |
| Downstream | coiled-coil domain containing 94 | Ccdc94 |
2900016D05Rik, AI413813 |
2.2kb | Anti-sense | Chr. 17 - 55959187 - 55968285 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16712
Genomic Map