Assay Details
| Gene Symbol: Gm16835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147121.1 | - | 1 - 2 | 1109 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DnaJ (Hsp40) homolog, subfamily C, member 3 |
Dnajc3 |
AA408985, AU067833, Dnajc3a, Dnajc3b, Prkri, mp58, p58, p58IPK |
-117.3kb | Sense | Chr. 14 - 118937932 - 118981702 on NCBI Build GRCm38 |
| Downstream | heparan sulfate 6-O-sulfotransferase 3 | Hs6st3 |
6OST3, HS6ST-3 |
37.1kb | Sense | Chr. 14 - 119137660 - 119872871 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16835
Genomic Map