Assay Details
| Gene Symbol: Gm16835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147121.1 | - | 4 - 5 | 1640 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UDP-glucose glycoprotein glucosyltransferase 2 | Uggt2 |
1810064L21Rik, 3110001A05Rik, 3110027P15Rik, A230065J02Rik, AW047562, Ugcgl2 |
-6.7kb | Anti-sense | Chr. 14 - 118981765 - 119099437 on NCBI Build GRCm38 |
| Downstream | heparan sulfate 6-O-sulfotransferase 3 | Hs6st3 |
6OST3, HS6ST-3 |
27.3kb | Sense | Chr. 14 - 119137660 - 119872871 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16835
Genomic Map