Assay Details
| Gene Symbol: Gm31583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK149396.1 | - | 1 - 2 | 224 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | B cell leukemia/lymphoma 6 | Bcl6 | Bcl5 | -100.1kb | Sense | Chr. 16 - 23965052 - 23988612 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 38456 |
Gm38456 | 181.1kb | Sense | Chr. 16 - 24271143 - 24296233 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31583
Genomic Map