Assay Details
| Gene Symbol: Gm30505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110508.1 | 1 - 2 | 86 | 100 | ||
| GenBank mRNA | AK149453.1 | - | 1 - 2 | 88 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | alpha-2-HS-glycoprotein | Ahsg | Anti-sense | Chr. 16 - 22892015 - 22899451 on NCBI Build GRCm38 | ||
| Downstream | fetuin beta | Fetub |
2310011O17Rik, AI255764, D17980, Gugu |
24.0kb | Anti-sense | Chr. 16 - 22918382 - 22939768 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm30505
Genomic Map