Assay Details
| Gene Symbol: Gm20513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK153671.1 | - | 2 - 3 | 259 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
histocompatibility 2, class II antigen A, alpha |
H2-Aa |
Aalpha, H-2Aa, H2Aa, I-Aalpha, IAalpha, Ia-1, Ia1 |
-5.3kb | Sense | Chr. 17 - 34282751 - 34287771 on NCBI Build GRCm38 |
| Downstream |
histocompatibility 2, class II antigen E beta |
H2-Eb1 |
H-2Eb, H2Eb, Ia-4, Ia4 |
830.0 | Anti-sense | Chr. 17 - 34305867 - 34316674 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm20513
Genomic Map