Assay Details
| Gene Symbol: Gm15708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040432.1 | 2 - 3 | 309 | 96 | ||
| GenBank mRNA | AK153727.1 | - | 2 - 3 | 310 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | brain protein I3 | Bri3 | I3 | Sense | Chr. 5 - 144244437 - 144446757 on NCBI Build GRCm38 | |
| Downstream | Dmrt1 related | Dmr | 77.4kb | Sense | Chr. 5 - 144357923 - 144361005 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15708
Genomic Map