Assay Details
| Gene Symbol: Gm31169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK156678.1 | - | 7 - 8 | 1382 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C2 calcium-dependent domain containing 5 | C2cd5 |
5730419I09Rik, C030008B15Rik, CDP138 |
-9.2kb | Sense | Chr. 6 - 143010920 - 143100603 on NCBI Build GRCm38 |
| Downstream | ethanolamine kinase 1 | Etnk1 |
1110061E11Rik, 4930555L11Rik, AI195356, AI841245, C80956, D6Ertd3e, EKI1 |
33.3kb | Anti-sense | Chr. 6 - 143166866 - 143208546 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31169
Genomic Map