Assay Details
| Gene Symbol: Gm35387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC066010.1 | - | 1 - 2 | 564 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
calcium channel, voltage-dependent, alpha2/delta subunit 3 |
Cacna2d3 | Cacnad3 | -181.9kb | Anti-sense | Chr. 14 - 28904943 - 29721864 on NCBI Build GRCm38 |
| Downstream | selenoprotein K | Selk |
1110001C03Rik, AA673253, HSPC030, Hsp30 |
60.8kb | Sense | Chr. 14 - 29968380 - 29975074 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm35387
Genomic Map