Assay Details
| Gene Symbol: Gm16758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC066032.1 | - | 492 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAM domain containing glycosylphosphatidylinositol anchor 1 | Mdga1 |
1200011I03Rik, GPIM, Mamdc3 |
-1.7kb | Anti-sense | Chr. 17 - 29827956 - 29888429 on NCBI Build GRCm38 |
| Downstream |
zinc finger, AN1-type domain 3 |
Zfand3 |
AW539211, TEG-27, Tex27 |
113.9kb | Sense | Chr. 17 - 30005039 - 30210020 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16758
Genomic Map