Assay Details
| Gene Symbol: LOC102640042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC072619.1 | - | 1 - 2 | 48 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ladybird homeobox homolog 1 (Drosophila) | Lbx1 | Lbx1h | -98.0kb | Sense | Chr. 19 - 45232684 - 45237435 on NCBI Build GRCm38 |
| Downstream | beta-transducin repeat containing protein | Btrc |
Beta-Trcp1, E3RS-IkappaB, E3RSIkappaB, FWD1, Fbw1a, HOS, Slimb, b-TrCP, beta-TrCP |
25.4kb | Anti-sense | Chr. 19 - 45363714 - 45533343 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102640042
Genomic Map