Assay Details
| Gene Symbol: Gm32618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC089548.1 | - | 456 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 41289 |
Gm41289 | -20.4kb | Anti-sense | Chr. 15 - 32754690 - 32773152 on NCBI Build GRCm38 | |
| Downstream | syndecan 2 | Sdc2 |
4833414L08Rik, AA960457, Hspg1, Synd2, syndecan-2 |
125.4kb | Sense | Chr. 15 - 32920723 - 33034721 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm32618
Genomic Map