Assay Details
| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028593.1 | 3 - 4 | 965 | 85 | ||
| GenBank mRNA | BC091771.1 | - | 3 - 4 | 383 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FK506 binding protein 4 | Fkbp4 |
59kDa, AL022792, AW208983, FKBP-4, FKBP-52, FKBP52, FKPB52, p59 |
-50.3kb | Anti-sense | Chr. 6 - 128430107 - 128438631 on NCBI Build GRCm38 |
| Downstream | cDNA sequence BC048546 | BC048546 | Ovos | 37.5kb | Anti-sense | Chr. 6 - 128539822 - 128581606 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10069
Genomic Map