Assay Details
| Gene Symbol: Gm19939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045942.1 | - | 1 | 155 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | proline rich 16 | Prr16 |
5430406M13Rik, AI607429 |
Sense | Chr. 18 - 51117898 - 51305129 on NCBI Build GRCm38 | |
| Downstream | ferritin mitochondrial | Ftmt |
4930447C24Rik, FerH, Fth3, MtF |
1.2mb | Sense | Chr. 18 - 52331536 - 52332996 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm19939
Genomic Map