Assay Details
| Gene Symbol: Gm12916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079012.1 | - | 3 - 4 | 487 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Ras-related GTP binding C | Rragc |
AU041672, Gtr2, RAGC, TIB929, YGR163W |
-356.7kb | Anti-sense | Chr. 4 - 123917433 - 123936997 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700021L23 gene | 1700021L23Rik | 6.4kb | Sense | Chr. 4 - 124302835 - 124332720 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12916
Genomic Map