Assay Details
| Gene Symbol: Gm15239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080343.1 | - | 1 - 2 | 217 | 87 | |
| BB236368.2 | - | 1 - 2 | 240 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | toll-like receptor 7 | Tlr7 | -30.9kb | Anti-sense | Chr. X - 167304926 - 167330574 on NCBI Build GRCm38 | |
| Downstream | FERM and PDZ domain containing 4 | Frmpd4 |
Gm196, PKAP1, Pdzd10, Pdzk10, Preso1, preso |
106.9kb | Anti-sense | Chr. X - 167471301 - 168577244 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15239
Genomic Map