Assay Details
| Gene Symbol: Gm34220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK081464.1 | - | 1 | 1244 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
degenerative spermatocyte homolog 2 (Drosophila), lipid desaturase |
Degs2 |
2210008A03Rik, AI852933, DES2 |
-87.7kb | Sense | Chr. 12 - 108686792 - 108702306 on NCBI Build GRCm38 |
| Downstream | YY1 transcription factor | Yy1 |
AW488674, NF-E1 |
526.0 | Anti-sense | Chr. 12 - 108793269 - 108816632 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm34220
Genomic Map