Assay Details
| Gene Symbol: Gm31135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082626.1 | - | 1 - 2 | 37 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31343 |
Gm31343 | -44.9kb | Anti-sense | Chr. 8 - 8377090 - 8391702 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31463 |
Gm31463 | 135.9kb | Anti-sense | Chr. 8 - 8576522 - 8580342 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31135
Genomic Map