Assay Details
| Gene Symbol: Gm32273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK085773.1 | - | 1 | 757 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho guanine nucleotide exchange factor (GEF) 15 | Arhgef15 |
D130071N09, D530030K12Rik, E5 |
-49.0 | Anti-sense | Chr. 11 - 68927611 - 68956934 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 25, member 35 |
Slc25a35 | 1810012H11Rik | 10.0kb | Sense | Chr. 11 - 68968108 - 68972516 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm32273
Genomic Map