Assay Details
| Gene Symbol: Gm26704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087598.1 | - | 1 | 2551 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | frizzled homolog 6 (Drosophila) | Fzd6 | Fz6 | Anti-sense | Chr. 15 - 39005817 - 39038194 on NCBI Build GRCm38 | |
| Downstream | collagen triple helix repeat containing 1 | Cthrc1 | 1110014B07Rik | 70.7kb | Anti-sense | Chr. 15 - 39076851 - 39087121 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm26704
Genomic Map