Assay Details
| Gene Symbol: Gm41829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089320.1 | - | 1 | 2147 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNA terminal phosphate cyclase-like 1 | Rcl1 |
2310040A02Rik, AI789745, C76567, RPCL1, Rnac |
-95.6kb | Anti-sense | Chr. 19 - 29101375 - 29143851 on NCBI Build GRCm38 |
| Downstream | Janus kinase 2 | Jak2 | Fd17 | 10.1kb | Anti-sense | Chr. 19 - 29251776 - 29313080 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm41829
Genomic Map