Assay Details
| Gene Symbol: Gm34433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC044776.2 | - | 2 - 3 | 143 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FBJ osteosarcoma oncogene B | Fosb | -26.6kb | Anti-sense | Chr. 7 - 19302696 - 19310226 on NCBI Build GRCm38 | |
| Downstream |
excision repair cross-complementing rodent repair deficiency, complementation group 1 |
Ercc1 | Ercc-1 | 6.8kb | Sense | Chr. 7 - 19344422 - 19356524 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm34433
Genomic Map