Assay Details
| Gene Symbol: Gm32854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC048761.1 | - | 1 - 2 | 241 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
calcium channel, voltage-dependent, P/Q type, alpha 1A subunit |
Cacna1a |
APCA, BI, Caca1a, Cacnl1a4, Cav2.1, Ccha1a, EA2, FHM, HPCA, MHP, MHP1, SCA6, alpha1A, la, nmf352, rkr, tg |
Anti-sense | Chr. 8 - 84338633 - 84640249 on NCBI Build GRCm38 | |
| Downstream | immediate early response 2 | Ier2 |
AI317238, Ch1, Pip92 |
73.7kb | Sense | Chr. 8 - 84661331 - 84662852 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm32854
Genomic Map