Assay Details
Gene Symbol: Gm30498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110507.1 | 4 - 5 | 275 | 101 | ||
GenBank mRNA | BC049705.1 | - | 4 - 5 | 275 | 101 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BU939480.1 | - | 278 | Chr. - - [-] on Build | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700069P05 gene | 1700069P05Rik | -53.8kb | Sense | Chr. 6 - 118246695 - 118248454 on NCBI Build GRCm38 | |
Downstream |
BMS1 homolog, ribosome assembly protein (yeast) |
Bms1 |
AA408648, AU020092, AU043373, BB007109, BC030906, Bms1l, mKIAA0187 |
77.6kb | Sense | Chr. 6 - 118383381 - 118420018 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
