Assay Details
| Gene Symbol: Gm30498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110507.1 | 5 - 6 | 352 | 81 | ||
| GenBank mRNA | BC049705.1 | - | 5 - 6 | 352 | 81 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BU939480.1 | - | 355 | Chr. - - [-] on Build | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700069P05 gene | 1700069P05Rik | -51.7kb | Sense | Chr. 6 - 118246695 - 118248454 on NCBI Build GRCm38 | |
| Downstream |
BMS1 homolog, ribosome assembly protein (yeast) |
Bms1 |
AA408648, AU020092, AU043373, BB007109, BC030906, Bms1l, mKIAA0187 |
81.1kb | Sense | Chr. 6 - 118383381 - 118420018 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm30498
Genomic Map