Assay Details
| Gene Symbol: Gm20619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015224.1 | - | 3 - 4 | 298 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 13 |
Slc25a13 |
AI785475, Ctrn |
-98.6kb | Anti-sense | Chr. 6 - 6041218 - 6217233 on NCBI Build GRCm38 |
| Downstream | split hand/foot malformation (ectrodactyly) type 1 | Shfm1 |
DSS1, Shfdg1, Shfg |
179.6kb | Anti-sense | Chr. 6 - 6558275 - 6578658 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm20619
Genomic Map