Assay Details
Gene Symbol: Gm31912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK047239.1 | - | 490 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | hyaluronic acid binding protein 2 | Habp2 |
AI035669, FSAP, HABP, HGFAL, PHBP |
156.8kb | Sense | Chr. 19 - 56287113 - 56320089 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
