Assay Details
| Gene Symbol: Gm15396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131623.1 | - | 2 - 3 | 647 | 141 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
nuclear receptor subfamily 1, group H, member 2 |
Nr1h2 |
AI194859, LXR, LXRB, LXRbeta, NER1, OR-1, RIP15, UR, Unr, Unr2 |
-26.0kb | Sense | Chr. 7 - 44549616 - 44553969 on NCBI Build GRCm38 |
| Downstream |
potassium voltage gated channel, Shaw-related subfamily, member 3 |
Kcnc3 |
KShIIID, Kcr2-3, Kv3.3 |
9.1kb | Anti-sense | Chr. 7 - 44590661 - 44604754 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15396
Genomic Map