Assay Details
| Gene Symbol: Gm30127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131720.1 | - | 5 - 6 | 792 | 121 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribonuclease P 40 subunit | Rpp40 |
D8Bwg1265e, Rnasep1 |
-46.0kb | Sense | Chr. 13 - 35895104 - 35911565 on NCBI Build GRCm38 |
| Downstream | LYR motif containing 4 | Lyrm4 |
BC034664, Gm903 |
19.6kb | Sense | Chr. 13 - 35978797 - 36117357 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm30127
Genomic Map