Assay Details
| Gene Symbol: Gm29154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131742.1 | - | 1 - 2 | 126 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 108 | Tmem108 |
AI462967, B130017P16Rik, R74726 |
-177.6kb | Sense | Chr. 9 - 103482936 - 103761872 on NCBI Build GRCm38 |
| Downstream | nephronophthisis 3 (adolescent) | Nphp3 |
3632410F03Rik, AI550417, C230078J01, D330020E01Rik, pcy |
43.1kb | Anti-sense | Chr. 9 - 104002544 - 104043811 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29154
Genomic Map