Assay Details
| Gene Symbol: Gm13964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131757.1 | - | 2 - 3 | 488 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | formin 1 | Fmn1 |
Fmn, ld |
Anti-sense | Chr. 2 - 113327699 - 113716768 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930533B01 gene | 4930533B01Rik | 187.7kb | Sense | Chr. 2 - 113691053 - 113708550 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13964
Genomic Map