Assay Details
| Gene Symbol: Gm35191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135912.1 | - | 3 - 4 | 268 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heparan sulfate (glucosamine) 3-O-sulfotransferase 1 | Hs3st1 |
3-Ost, D5Wsu110e, Hsg3ost |
-331.0kb | Sense | Chr. 5 - 39613935 - 39644631 on NCBI Build GRCm38 |
| Downstream |
RAB28, member RAS oncogene family |
Rab28 |
2700023P08Rik, AW496496 |
1.6mb | Sense | Chr. 5 - 41624974 - 41708192 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm35191
Genomic Map