Assay Details
| Gene Symbol: Gm14936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK136133.1 | - | 3 - 4 | 380 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | premature ovarian failure 1B | Pof1b | 2310066B14Rik | -282.2kb | Sense | Chr. X - 112638427 - 112698711 on NCBI Build GRCm38 |
| Downstream | choroidermia (RAB escort protein 1) | Chm | Rep-1 | 48.8kb | Sense | Chr. X - 113040590 - 113185539 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm14936
Genomic Map