Assay Details
| Gene Symbol: Gm15201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK136312.1 | - | 1 - 2 | 242 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RALBP1 associated Eps domain containing protein 2 | Reps2 | POB1 | -91.4kb | Sense | Chr. X - 162411952 - 162643705 on NCBI Build GRCm38 |
| Downstream | retinoblastoma binding protein 7 | Rbbp7 |
AA409861, AI173248, AU019541, BB114024, mRbAp46 |
23.8kb | Anti-sense | Chr. X - 162760372 - 162779090 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15201
Genomic Map