Assay Details
| Gene Symbol: Gm31152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK141156.1 | - | 538 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 110 | Zfp110 |
2900024E01Rik, N28112, NRIF, Nrif1 |
-15.9kb | Anti-sense | Chr. 7 - 12834810 - 12850584 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 128 | Zfp128 |
9630016P15Rik, Znf8, mZnf8 |
14.5kb | Anti-sense | Chr. 7 - 12881178 - 12893422 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm31152
Genomic Map