Assay Details
| Gene Symbol: Gm34637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144171.1 | - | 1 - 2 | 87 | 121 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4921511I17 gene | 4921511I17Rik | -276.3kb | Sense | Chr. 12 - 12351522 - 12392475 on NCBI Build GRCm38 | |
| Downstream |
v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) |
Mycn |
N-myc, Nmyc, Nmyc-1, Nmyc1, bHLHe37, c-nmyc |
266.2kb | Sense | Chr. 12 - 12936093 - 12941836 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm34637
Genomic Map