Assay Details
| Gene Symbol: Gm13544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040365.1 | 2 - 3 | 292 | 85 | ||
| GenBank mRNA | AK144626.1 | - | 2 - 3 | 293 | 85 | |
| AK165961.1 | - | 2 - 3 | 292 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
activin A receptor, type IC |
Acvr1c |
ACVRLK7, ALK7, Alk-7, C230097P10 |
Anti-sense | Chr. 2 - 58267453 - 58357896 on NCBI Build GRCm38 | |
| Downstream |
activin A receptor, type 1 |
Acvr1 |
ALK2, ActR-I, ActRIA, Acvr, Acvrlk2, Alk-2, Alk8, D330013D15Rik, SKR1, Tsk7L |
165.1kb | Anti-sense | Chr. 2 - 58446438 - 58566828 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0300000629.rn
Set Membership: |
|

Gm13544
Genomic Map