Assay Details
| Gene Symbol: Gm32282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144664.1 | - | 1 - 2 | 432 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4710 | Gm4710 | -2.3mb | Anti-sense | Chr. 17 - 75861023 - 75889039 on NCBI Build GRCm38 | |
| Downstream | cysteine rich transmembrane BMP regulator 1 (chordin like) | Crim1 | AU015004 | 160.0 | Anti-sense | Chr. 17 - 78200185 - 78376592 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm32282
Genomic Map