Assay Details
| Gene Symbol: Gm15834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053027.1 | - | 2 - 3 | 1767 | 72 | |
| AK144771.1 | - | 3 - 4 | 274 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
origin recognition complex, subunit 2 |
Orc2 |
AU041563, Orc2l |
-7.0kb | Anti-sense | Chr. 1 - 58462771 - 58505109 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 126, member B |
Fam126b |
C130065N10Rik, D1Ertd53e, D630010C10 |
5.3kb | Anti-sense | Chr. 1 - 58522806 - 58586333 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15834
Genomic Map