Assay Details
Gene Symbol: Gm15473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144868.1 | - | 1 - 2 | 411 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transcription factor EC | Tfec |
BB107417, Tcfec, bHLHe34 |
-160.3kb | Sense | Chr. 6 - 16830837 - 16898462 on NCBI Build GRCm38 |
Downstream | testis derived transcript | Tes |
D6Ertd352e, TESS, Tes1, Tes2, testin2 |
153.0 | Anti-sense | Chr. 6 - 17065149 - 17105825 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
