Assay Details
| Gene Symbol: Gm16845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040406.1 | 3 - 4 | 333 | 88 | ||
| NR_040407.1 | 2 - 3 | 269 | 88 | |||
| GenBank mRNA | AA021721.1 | - | 3 - 4 | 333 | 88 | |
| AI509114.1 | - | 3 - 4 | 333 | 88 | ||
| AK154427.1 | - | 2 - 3 | 239 | 88 | ||
| BY232108.1 | - | 2 - 3 | 255 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ECSIT homolog (Drosophila) | Ecsit | Sitpec | Anti-sense | Chr. 9 - 22071108 - 22085443 on NCBI Build GRCm38 | |
| Downstream | calponin 1 | Cnn1 |
CN, CnnI |
13.1kb | Sense | Chr. 9 - 22099221 - 22109221 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16845
Genomic Map