Assay Details
| Gene Symbol: Gm35363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK158044.1 | - | 2 - 3 | 361 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphodiesterase 2A, cGMP-stimulated |
Pde2a |
CGS-PDE, cGSPDE |
-13.6kb | Anti-sense | Chr. 7 - 101421691 - 101512829 on NCBI Build GRCm38 |
| Downstream |
ADP-ribosyltransferase 2a, pseudogene |
Art2a-ps |
ARTC2, Art2a, Ly92a, Rt-6, Rt6, Rt6-1 |
20.1kb | Sense | Chr. 7 - 101552453 - 101560865 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm35363
Genomic Map