Assay Details
| Gene Symbol: Gm12348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK158747.1 | - | 3 - 4 | 362 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | junction plakoglobin | Jup |
Ctnng, D930025P04Rik, PG |
-4.3kb | Anti-sense | Chr. 11 - 100368856 - 100397790 on NCBI Build GRCm38 |
| Downstream | prolyl 3-hydroxylase family member 4 (non-enzymatic) | P3h4 |
1110036O03Rik, AI413214, Leprel4, Sc65 |
4.9kb | Anti-sense | Chr. 11 - 100408748 - 100414819 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12348
Genomic Map